5,6-Dibromo-2-pyridinamine 97%
Lieferant: Apollo Scientific
Synonyme:
5,6-Dibromopyridin-2-amine
OR301215-5G
APOSOR301215-5GEA
140
EUR
APOSOR301215-5G
5,6-Dibromo-2-pyridinamine 97%
5,6-Dibromo-2-pyridinamine
Formel:
C₅H₄Br₂N₂ Molecular Weight: 251.907 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234045 CAS-Nummer: 89284-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...