3-Chloro-2-iodoaniline 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR301216-5G
APOSOR301216-5GEA
87
EUR
APOSOR301216-5G
3-Chloro-2-iodoaniline 98%
3-Chloro-2-iodoaniline
Formel:
C₆H₅ClIN Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738966 CAS-Nummer: 70237-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...