4-Benzylmorpholine-2-carbonitrile

Lieferant: Apollo Scientific

OR301246-1G
APOSOR301246-1GEA 0 EUR
APOSOR301246-1G
4-Benzylmorpholine-2-carbonitrile
4-Benzylmorpholine-2-carbonitrile
Formel: C₁₂H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08751344
CAS-Nummer: 126645-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR