1H-Imidazole-1-sulphonyl azide hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR301255-1G
APOSOR301255-1GEA 308 EUR
APOSOR301255-1G
1H-Imidazole-1-sulphonyl azide hydrochloride
1H-Imidazole-1-sulphonyl azide hydrochloride
Formel: C₃H₃N₅O₂S·ClH
Molecular Weight: 209,614 g/mol
Schmelzpunkt: 99…104 °C
Lagertemperatur: Kühlschrank
MDL: MFCD19705431
CAS-Nummer: 952234-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR