6-Chloro-1H-pyrrolo[3,2-c]pyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-5-azaindole

OR301258-5G
APOSOR301258-5GEA 101 EUR
APOSOR301258-5G
6-Chloro-1H-pyrrolo[3,2-c]pyridine 97%
6-Chloro-1H-pyrrolo[3,2-c]pyridine
Formel: C₇H₅ClN₂
Molecular Weight: 152,5800 g/mol
Schmelzpunkt: 190…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08690133
CAS-Nummer: 74976-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR