6-Bromo-1H-pyrrolo[3,2-b]pyridine 97%

Lieferant: Apollo Scientific

OR301260-25G
APOSOR301260-25GEA 137 EUR
APOSOR301260-25G
6-Bromo-1H-pyrrolo[3,2-b]pyridine 97%
6-Bromo-1H-pyrrolo[3,2-b]pyridine
Formel: C₇H₅BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09266227
CAS-Nummer: 944937-53-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR