1H-Pyrrolo[2,3-b]pyridin-4-amine hydrochloride 97%

Lieferant: Apollo Scientific

OR301265-1G
APOSOR301265-1GEA 125 EUR
APOSOR301265-1G
1H-Pyrrolo[2,3-b]pyridin-4-amine hydrochloride 97%
1H-Pyrrolo[2,3-b]pyridin-4-amine hydrochloride
Formel: C₇H₇N₃·ClH
Molecular Weight: 169,612 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689965
CAS-Nummer: 1134307-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR