5-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Bromo-3-iodo-7-azaindole
OR301271-1G
OR301271-5G
APOSOR301271-1GEA
21.9
EUR
APOSOR301271-1G
APOSOR301271-5G
5-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine 95%
5-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine
Formel:
C₇H₄BrIN₂ Schmelzpunkt: 234…239 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779519 CAS-Nummer: 757978-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...