5-Bromo-2-methyl-7-azaindole
Lieferant: Apollo Scientific
|
Achtung
|
OR301272-1G
APOSOR301272-1GEA
89.9
EUR
APOSOR301272-1G
5-Bromo-2-methyl-7-azaindole
5-Bromo-2-methyl-7-azaindole
Formel:
C₈H₇BrN₂ Molecular Weight: 211,0600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15529124 CAS-Nummer: 1111638-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...