6-Chloro-1H-pyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 6-Chloro-7-azaindole

OR301275-25G
APOSOR301275-25GEA 161 EUR
APOSOR301275-25G
6-Chloro-1H-pyrrolo[2,3-b]pyridine
6-Chloro-1H-pyrrolo[2,3-b]pyridine
Formel: C₇H₅ClN₂
Molecular Weight: 152.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659666
CAS-Nummer: 55052-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR