Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 1H-pyrrolo[2,3-b]pyridine-5-carboxylate

OR301280-5G
APOSOR301280-5GEA 139 EUR
APOSOR301280-5G
Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%
Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate
Formel: C₉H₈N₂O₂
Molecular Weight: 176.1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08457785
CAS-Nummer: 849067-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR