5-Chlor-2-methoxybenzonitril
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Chlor-2-methoxybenzoesäurenitril
OR30129-250MG
APOSOR30129-250MGEA
21.9
EUR
APOSOR30129-250MG
5-Chlor-2-methoxybenzonitril
5-Chlor-2-methoxybenzonitril
Formel:
C₈H₆ClNO Molecular Weight: 167,59 g/mol Schmelzpunkt: 98…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052867 CAS-Nummer: 55877-79-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...