1-Boc-2-(aminomethyl)-piperidin

Lieferant: Apollo Scientific

OR301292-1G
APOSOR301292-1GEA 0 EUR
APOSOR301292-1G
1-Boc-2-(aminomethyl)-piperidin
1-Boc-(2-aminomethyl)piperidine
Formel: C₁₁H₂₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01317791
CAS-Nummer: 370069-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR