Ethyl 6-(aminomethyl)nicotinate hydrochloride 95+%

Lieferant: Apollo Scientific

OR301304-250MG
APOSOR301304-250MGEA 49.3 EUR
APOSOR301304-250MG
Ethyl 6-(aminomethyl)nicotinate hydrochloride 95+%
Ethyl 6-(aminomethyl)nicotinate hydrochloride
Formel: C₉H₁₂N₂O₂·ClH
Molecular Weight: 216,665 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09788628
CAS-Nummer: 1189434-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR