7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 98%

Lieferant: Apollo Scientific

OR301310-250MG
APOSOR301310-250MGEA 92.8 EUR
APOSOR301310-250MG
7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 98%
7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde
Formel: C₇H₅N₃O
Molecular Weight: 147,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189361
CAS-Nummer: 1060815-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR