7-Hydroxy-1-indanone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Hydroxy-2,3-dihydro-1H-inden-1-one
OR301315-5G
APOSOR301315-5GEA
128
EUR
APOSOR301315-5G
7-Hydroxy-1-indanone
7-Hydroxy-1-indanone
Formel:
C₉H₈O₂ Molecular Weight: 148.161 g/mol Siedepunkt: 131 °C/9.5mmHg Schmelzpunkt: 109…113 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01548387 CAS-Nummer: 6968-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...