3-Amino-7-bromisochinolin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Bromoisoquinolin-3-amine
OR301335-100MG
APOSOR301335-100MGEA
48
EUR
APOSOR301335-100MG
3-Amino-7-bromisochinolin
3-Amino-7-bromoisoquinoline
Formel:
C₉H₇BrN₂ Molecular Weight: 223.0693 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040207 CAS-Nummer: 1192815-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...