1-Oxa-7-azaspiro[3.5]nonane hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Oxa-7-azaspiro[3.5]nonane
OR301345-1G
OR301345-250MG
APOSOR301345-1GEA
493
EUR
APOSOR301345-1G
APOSOR301345-250MG
1-Oxa-7-azaspiro[3.5]nonane hydrochloride
1-Oxa-7-azaspiro[3.5]nonane hydrochloride
Formel:
C₇H₁₄ClNO Molecular Weight: 163.65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28139317 CAS-Nummer: 38674-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...