3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, 3-{[(tert-butoxy)carbonyl]amino}cyclobutane-1-carboxylic acid
OR301346-1G
APOSOR301346-1GEA
79.9
EUR
APOSOR301346-1G
3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid 97%
3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid
Formel:
C₁₀H₁₇NO₄ Molecular Weight: 215,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864181 CAS-Nummer: 946152-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...