Dihydro-2H-pyran-3(4H)-one 95+%

Lieferant: Apollo Scientific
Achtung

OR301349-1G
APOSOR301349-1GEA 23.2 EUR
APOSOR301349-1G
Dihydro-2H-pyran-3(4H)-one 95+%
Dihydro-2H-pyran-3(4H)-one
Formel: C₅H₈O₂
Molecular Weight: 100,1158 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00182426
CAS-Nummer: 23462-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR