3-(Cyclopentyloxy)benzenemethanamine hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: (3-(Cyclopentyloxy)phenyl)methanamine hydrochloride

OR301360-1G
APOSOR301360-1GEA 79.9 EUR
APOSOR301360-1G
3-(Cyclopentyloxy)benzenemethanamine hydrochloride 98%
3-(Cyclopentyloxy)benzenemethanamine hydrochloride
Formel: C₁₂H₁₇NO·ClH
Molecular Weight: 227,73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16040134
CAS-Nummer: 1235441-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR