1,3-Dichlor-2-iodbenzol 98%

Lieferant: Apollo Scientific
Achtung

OR301367-100G
APOSOR301367-100GEA 84.1 EUR
APOSOR301367-100G
1,3-Dichlor-2-iodbenzol 98%
1,3-Dichloro-2-iodobenzene
Formel: C₆H₃Cl₂I
Molecular Weight: 272,90 g/mol
Schmelzpunkt: 64…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060658
CAS-Nummer: 19230-28-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR