3-Amino-N-(tert-butoxycarbonyl)benzylamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: alpha-(Boc-amino)-m-toluidine, 3-Amino-N-Boc-benzylamine, tert-Butyl (3-Aminobenzyl)carbamate, alpha-(tert-Butoxycarbonyl)amino-m-toluidine, 3-[(tert-Butoxycarbonylamino)methyl]aniline, 3-[(Boc-amino)methyl]aniline, (3-Aminobenzyl)carbamic Acid tert-Butyl, tert-Butyl 3-aminobenzylcarbamate

OR301370-1G
APOSOR301370-1GEA 84.1 EUR
APOSOR301370-1G
3-Amino-N-(tert-butoxycarbonyl)benzylamine 97%
3-Amino-N-(tert-butoxycarbonyl)benzylamine
Formel: C₁₂H₁₈N₂O₂
Molecular Weight: 222,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04974010
CAS-Nummer: 147291-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR