Methyl-2-(brommethyl)benzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-bromomethylbenzoate

OR301372-25G
APOSOR301372-25GEA 112 EUR
APOSOR301372-25G
Methyl-2-(brommethyl)benzoat
Methyl-2-(brommethyl)benzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229,0706 g/mol
Schmelzpunkt: 32…33 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425900
CAS-Nummer: 2417-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR