2-(Boc-amino)-4-chlorpyridin

Lieferant: Apollo Scientific

OR301375-1G
APOSOR301375-1GEA 0 EUR
APOSOR301375-1G
2-(Boc-amino)-4-chlorpyridin
2-(Boc-amino)-4-chlorpyridin
Formel: C₁₀H₁₃ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05663519
CAS-Nummer: 130721-78-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR