2-Chloro-5-pyridinesulphonyl chloride 97%
Lieferant: Apollo Scientific
OR301377-5G
APOSOR301377-5GEA
85.7
EUR
APOSOR301377-5G
2-Chloro-5-pyridinesulphonyl chloride 97%
2-Chloro-5-pyridinesulphonyl chloride
Formel:
C₅H₃Cl₂NO₂S Molecular Weight: 212,0538 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03541049 CAS-Nummer: 6684-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...