2,3-Dibromopyrazine 95%

Lieferant: Apollo Scientific
Achtung

OR301382-1G
APOSOR301382-1GEA 79.9 EUR
APOSOR301382-1G
2,3-Dibromopyrazine 95%
2,3-Dibromopyrazine
Formel: C₄H₂Br₂N₂
Molecular Weight: 237,8800 g/mol
Schmelzpunkt: 57…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02684897
CAS-Nummer: 95538-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR