2,5-Dichlor-3-formylpyridin 95%
Lieferant: Apollo Scientific
Synonyme:
2,5-Dichloronicotinaldehyde, 2,5-Dichloro-3-formylpyridine
OR301386-5G
APOSOR301386-5GEA
0
EUR
APOSOR301386-5G
2,5-Dichlor-3-formylpyridin 95%
2,5-Dichlor-3-formylpyridin
Formel:
C₆H₃Cl₂NO Molecular Weight: 176,0000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08056627 CAS-Nummer: 176433-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...