5-Chlor-2-methoxybenzolsulfonylchlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Chlor-2-methoxybenzolsulfochlorid, 5-Chlor-2-methoxybenzolsulfonsäurechlorid
OR30141-25G
APOSOR30141-25GEA
75.4
EUR
APOSOR30141-25G
5-Chlor-2-methoxybenzolsulfonylchlorid 97%
5-Chlor-2-methoxybenzolsulfonylchlorid
Formel:
C₇H₆Cl₂O₃S Molecular Weight: 241.09 g/mol Schmelzpunkt: 100…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052959 CAS-Nummer: 22952-32-5 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...