Methyl-4-chlorbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chlorbenzoesäuremethylester

OR30145-500G
APOSOR30145-500GEA 159 EUR
APOSOR30145-500G
Methyl-4-chlorbenzoat
Methyl-4-chlorbenzoat
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000621
CAS-Nummer: 1126-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR