methyl 4-aminobenzoate hydrochloride

Lieferant: Apollo Scientific

OR30151-1G OR30151-250MG OR30151-500MG
APOSOR30151-1GEA 0 EUR
APOSOR30151-1G APOSOR30151-250MG APOSOR30151-500MG
methyl 4-aminobenzoate hydrochloride
methyl 4-aminobenzoate hydrochloride
Formel: C₈H₉NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD00205145
CAS-Nummer: 63450-84-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR