4-Phenyl-5-thio-3H-1,2-dithiole-3-thione

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Mercapto-4-phenyl-3H-1,2-dithiole-3-thione, 4-Phenyl-5-sulphanyl-3H-1,2-dithiole-3-thione, 3-Oxo-4-phenyl-5-sulphanyl-3H-1,2-dithiole

OR30153-1G
APOSOR30153-1GEA 91.5 EUR
APOSOR30153-1G
4-Phenyl-5-thio-3H-1,2-dithiole-3-thione
4-Phenyl-5-thio-3H-1,2-dithiole-3-thione
Formel: C₉H₆S₄
Molecular Weight: 242,404 g/mol
Schmelzpunkt: 149…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052322
CAS-Nummer: 16101-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR