(4-Bromphenyl)essigsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR30163-100G
OR30163-25G
APOSOR30163-100GEA
29
EUR
APOSOR30163-100G
APOSOR30163-25G
(4-Bromphenyl)essigsäure 97%
(4-Bromphenyl)essigsäure
Formel:
(C₆H₄Br)CH₂COOH Schmelzpunkt: 117…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004342 CAS-Nummer: 1878-68-8 EINECS: 217-523-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...