4-Amino-2-chlorpyrimidin-5-carbosäurenitril
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Amino-2-chloro-5-pyrimidinecarbonitrile
OR30167-1G
APOSOR30167-1GEA
21.9
EUR
APOSOR30167-1G
4-Amino-2-chlorpyrimidin-5-carbosäurenitril
4-Amino-2-chlorpyrimidin-5-carbosäurenitril
Formel:
C₅H₃ClN₄ Molecular Weight: 154,56 g/mol Schmelzpunkt: 243…244 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052343 CAS-Nummer: 94741-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...