Furil 97%

Lieferant: Apollo Scientific

Synonyme: 1,2-di(2-furyl)ethane-1,2-dione, 1,2-Di(furan-2-yl)ethane-1,2-dione

OR30173-25G
APOSOR30173-25GEA 241 EUR
APOSOR30173-25G
Furil 97%
Furil
Formel: C₁₀H₆O₄
Molecular Weight: 190,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003241
CAS-Nummer: 492-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR