1-Methyl-4-nitro-1H-pyrazole 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-4-nitropyrazole

OR302024-100G
APOSOR302024-100GEA 74.1 EUR
APOSOR302024-100G
1-Methyl-4-nitro-1H-pyrazole 98%
1-Methyl-4-nitro-1H-pyrazole
Formel: C₄H₅N₃O₂
Molecular Weight: 127.10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00464261
CAS-Nummer: 3994-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR