3H-Imidazo[4,5-c]pyridine 5-oxide
Lieferant: Apollo Scientific
|
Achtung
|
OR302028-100MG
APOSOR302028-100MGEA
244
EUR
APOSOR302028-100MG
3H-Imidazo[4,5-c]pyridine 5-oxide
3H-Imidazo[4,5-c]pyridine 5-oxide
Formel:
C₆H₅N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD22989413 CAS-Nummer: 91184-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...