2-Amino-4-methylbenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR30203-5G
APOSOR30203-5GEA
21.9
EUR
APOSOR30203-5G
2-Amino-4-methylbenzoic acid
2-Amino-4-methylbenzoic acid
Formel:
C₈H₉NO₂ Schmelzpunkt: 174…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047853 CAS-Nummer: 2305-36-4 EINECS: 218-976-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...