1-Benzyl-4,5-dioxopyrrolidine-3-carboxylic acid ethyl ester
Lieferant: Apollo Scientific
Synonyme:
Ethyl 1-benzyl-4,5-dioxopyrrolidine-3-carboxylate
OR302032-1G
APOSOR302032-1GEA
0
EUR
APOSOR302032-1G
1-Benzyl-4,5-dioxopyrrolidine-3-carboxylic acid ethyl ester
1-Benzyl-4,5-dioxopyrrolidine-3-carboxylic acid ethyl ester
Formel:
C₁₄H₁₅NO₄ Molecular Weight: 261,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00098219 CAS-Nummer: 5336-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...