3,5-Dichloro-2-nitroaniline

Lieferant: Apollo Scientific

OR302035-1G
APOSOR302035-1GEA 221 EUR
APOSOR302035-1G
3,5-Dichloro-2-nitroaniline
3,5-Dichloro-2-nitroaniline
Formel: C₆H₄Cl₂N₂O₂
Molecular Weight: 207,014 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01463654
CAS-Nummer: 122584-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR