Furo[2,3-c]pyridine-2-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Carboxy-6-azabenzo[b]furan, 2-Carboxyfuro[2,3-c]pyridine
OR302052-1G
APOSOR302052-1GEA
1210
EUR
APOSOR302052-1G
Furo[2,3-c]pyridine-2-carboxylic acid 95%
Furo[2,3-c]pyridine-2-carboxylic acid
Formel:
C₈H₅NO₃ Molecular Weight: 163,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11869932 CAS-Nummer: 112372-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...