2-Methylbutylisothiocyanat

Lieferant: Apollo Scientific
Gefahr

OR30206-5G
APOSOR30206-5GEA 161 EUR
APOSOR30206-5G
2-Methylbutylisothiocyanat
2-Methylbutylisothiocyanat
Formel: C₆H₁₁NS
Molecular Weight: 129.23 g/mol
Siedepunkt: 84…86 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00060414
CAS-Nummer: 4404-51-7
UN: 2927
ADR: 6.1+8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR