N-Methyl-N-4-piperidinylmethanesulphonamide

Lieferant: Apollo Scientific

Synonyme: N-Methyl-N-(piperidin-4-yl)methanesulphonamide

OR302060-1G
APOSOR302060-1GEA 388 EUR
APOSOR302060-1G
N-Methyl-N-4-piperidinylmethanesulphonamide
N-Methyl-N-4-piperidinylmethanesulphonamide
Formel: C₇H₁₆N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD12086666
CAS-Nummer: 70724-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR