4-Cyclohexyl-3-thiosemicarbazid

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Cyclohexylhydrazinecarbothioamide

OR302062-5G
APOSOR302062-5GEA 215 EUR
APOSOR302062-5G
4-Cyclohexyl-3-thiosemicarbazid
4-Cyclohexyl-3-thiosemicarbazid
Formel: C₇H₁₅N₃S
Molecular Weight: 173,2791 g/mol
Schmelzpunkt: 142…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00021318
CAS-Nummer: 21198-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR