2-Bromopyridine-3,4-diamine

Lieferant: Apollo Scientific
Achtung

OR302077-1G
APOSOR302077-1GEA 183 EUR
APOSOR302077-1G
2-Bromopyridine-3,4-diamine
2-Bromopyridine-3,4-diamine
Formel: C₅H₆BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12910485
CAS-Nummer: 189230-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR