1-(4-Methoxybenzyl)-4-bromo-5-nitro-1H-pyrazole
Lieferant: Apollo Scientific
OR302080-1G
APOSOR302080-1GEA
511
EUR
APOSOR302080-1G
1-(4-Methoxybenzyl)-4-bromo-5-nitro-1H-pyrazole
1-(4-Methoxybenzyl)-4-bromo-5-nitro-1H-pyrazole
Formel:
C₁₁H₁₀BrN₃O₃ Molecular Weight: 312,119 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22690743 CAS-Nummer: 1429309-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...