2,4-Dichloro-7,7-dimethyl-5,7-dihydrofuro[3,4-d]pyrimidine

Lieferant: Apollo Scientific

OR302091-1G
APOSOR302091-1GEA 1200 EUR
APOSOR302091-1G
2,4-Dichloro-7,7-dimethyl-5,7-dihydrofuro[3,4-d]pyrimidine
2,4-Dichloro-7,7-dimethyl-5,7-dihydrofuro[3,4-d]pyrimidine
Formel: C₈H₈Cl₂N₂O
Molecular Weight: 219,068 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19688042
CAS-Nummer: 1260088-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR