4-Bromo-3-nitro-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR302098-1G
APOSOR302098-1GEA 27.7 EUR
APOSOR302098-1G
4-Bromo-3-nitro-1H-pyrazole
4-Bromo-3-nitro-1H-pyrazole
Formel: C₃H₂BrN₃O₂
Molecular Weight: 191.971 g/mol
Schmelzpunkt: 200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01114992
CAS-Nummer: 89717-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR