4-(4-Bromopyridin-2-yl)piperazine, N1-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Bromopyridin-2-yl)-4-(tert-butoxycarbonyl)piperazine, tert-Butyl 4-(4-bromopyridin-2-yl)piperazine-1-carboxylate
OR302099-1G
APOSOR302099-1GEA
59.6
EUR
APOSOR302099-1G
4-(4-Bromopyridin-2-yl)piperazine, N1-BOC protected
4-(4-Bromopyridin-2-yl)piperazine, N1-BOC protected
Formel:
C₁₄H₂₀BrN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13195778 CAS-Nummer: 1197294-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...