5-Bromo-1-methyl-4-nitro-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR302101-1G
APOSOR302101-1GEA 194 EUR
APOSOR302101-1G
5-Bromo-1-methyl-4-nitro-1H-pyrazole
5-Bromo-1-methyl-4-nitro-1H-pyrazole
Formel: C₄H₄BrN₃O₂
Molecular Weight: 205,997 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00195044
CAS-Nummer: 89607-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR